3,3'-Thiodipropionic acid, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
3,3-Thiodipropionic acid
M-Clarity™ quality level = MQ200
T30201-100G
T30201-3KG
SIALT30201-100GEA
20.5
GBP
SIALT30201-100G
SIALT30201-3KG
3,3'-Thiodipropionic acid, Sigma-Aldrich®
3,3'-Thiodipropionic acid
Formula:
C₆H₁₀O₄S MW: 178.21 g/mol Boiling Pt: 347 °C (1013 hPa) Melting Pt: 128…131 °C Density: 1.496 g/cm³ (20 °C) |
MDL Number:
MFCD00002781 CAS Number: 111-17-1 EINECS: 203-841-3 Merck Index: 12,09468 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...