3-(Hydroxymethyl)indole, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
3-Indolmethanol, 3-Indolcarbinol, (1H-Indol-3-yl)methanol, Indole-3-methanol, (Indole-3-yl)methanol, 3-Indolemethanol, Indole-3-carbinol
M-Clarity™ quality level = MQ100
I7256-1G
I7256-25G
I7256-5G
SIALI7256-1GEA
45.5
GBP
SIALI7256-1G
SIALI7256-25G
SIALI7256-5G
3-(Hydroxymethyl)indole, Sigma-Aldrich®
3-(Hydroxymethyl)indole
Formula:
C₉H₉NO MW: 147.18 g/mol |
MDL Number:
MFCD00005632 CAS Number: 700-06-1 EINECS: 211-836-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...