3-(4-Hydroxyphenyl)propionic acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-(4-Hydroxyphenyl)propanoic acid, p-hydroxyl phenylpropanol, 3-(p-Hydroxyphenyl)-Propionic Acid

M-Clarity™ quality level = MQ200

H52406-25G H52406-50G H52406-5G
SIALH52406-25GEA 101 GBP
SIALH52406-25G SIALH52406-50G SIALH52406-5G
3-(4-Hydroxyphenyl)propionic acid, Sigma-Aldrich®
3-(4-Hydroxyphenyl)propionic acid
Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 208…210 °C (14 mmHg)
Melting Pt: 126…129 °C
Density: 1.336 g/cm³ (20 °C)
MDL Number: MFCD00002778
CAS Number: 501-97-3
EINECS: 207-931-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR