5-Amino-2-methoxypyridine, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ100
A61209-25G
A61209-5G
SIALA61209-25GEA
101
GBP
SIALA61209-25G
SIALA61209-5G
5-Amino-2-methoxypyridine, Sigma-Aldrich®
5-Amino-2-methoxypyridine
Formula:
C₆H₈N₂O MW: 124.14 g/mol Boiling Pt: 265 °C (1013 hPa) Melting Pt: 28…30 °C |
MDL Number:
MFCD00006264 CAS Number: 6628-77-9 EINECS: 229-612-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...