1-Butyl-3-methylimidazolium methyl sulphate, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
[BMIM] [MeSO4], 1-n-Butyl-3-methylimidazolium methyl sulphate, ECOENG 411, BMIMMeSO4
83086-5G
83086-50G
SIAL83086-5GEA
115
GBP
SIAL83086-5G
SIAL83086-50G
1-Butyl-3-methylimidazolium methyl sulphate, Sigma-Aldrich®
1-Butyl-3-methylimidazolium methyl sulphate
Formula:
C₉H₁₈N₂O₄S MW: 250.32 g/mol Density: 1.21 g/cm³ (20 °C) |
MDL Number:
MFCD03095437 CAS Number: 401788-98-5 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...