Pyridinium chloride, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
M-Clarity™ quality level = MQ200
82800-100G
82800-500G
SIAL82800-100GEA
88.2
GBP
SIAL82800-100G
SIAL82800-500G
Pyridinium chloride, Sigma-Aldrich®
Pyridinium chloride
Formula:
C₅H₆ClN Boiling Pt: 222.5 °C (1013 hPa) Melting Pt: 144 °C Density: 1.34 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00012802 CAS Number: 628-13-7 EINECS: 211-027-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...