Vinyltrimethylsilane, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Trimethyl(vinyl)silane

M-Clarity™ quality level = MQ100

SIAL661724-25GEA 539 GBP
SIAL661724-25G
Vinyltrimethylsilane, Sigma-Aldrich®
Vinyltrimethylsilane
Formula: C₅H₁₂Si
MW: 100.24 g/mol
Boiling Pt: 55 °C (1013 hPa)
Density: 0.7038 g/cm³ (20 °C)
MDL Number: MFCD00008606
CAS Number: 754-05-2
EINECS: 212-042-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR