3-(m-Tolyl)propionic acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-(3-Methylphenyl)propionic acid, 3-(3-Methylphenyl)propanoic acid

M-Clarity™ quality level = MQ100

656178-1G 656178-5G
SIAL656178-1GEA 58 GBP
SIAL656178-1G SIAL656178-5G
3-(m-Tolyl)propionic acid, Sigma-Aldrich®
3-(m-Tolyl)propionic acid
Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
MDL Number: MFCD00060326
CAS Number: 3751-48-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR