2-(Trifluoromethyl)pyridine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

643572-1G 643572-5G
SIAL643572-1GEA 51.9 GBP
SIAL643572-1G SIAL643572-5G
2-(Trifluoromethyl)pyridine, Sigma-Aldrich®
2-(Trifluoromethyl)pyridine
Formula: C₆H₄F₃N
MW: 147.1 g/mol
Boiling Pt: 138…139 °C (1013 hPa)
MDL Number: MFCD02183562
CAS Number: 368-48-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR