2-Chloro-3-(trifluoromethyl)pyridine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

469378-25G 469378-5G
SIAL469378-25GEA 99.6 GBP
SIAL469378-25G SIAL469378-5G
2-Chloro-3-(trifluoromethyl)pyridine, Sigma-Aldrich®
2-Chloro-3-(trifluoromethyl)pyridine
Formula: C₆H₃ClF₃N
MW: 181.54 g/mol
Boiling Pt: 75…76 °C (30 mmHg)
Melting Pt: 38…39 °C
MDL Number: MFCD00042223
CAS Number: 65753-47-1
EINECS: 424-520-6
UN: 2923
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR