α,α,α-Trifluoro-3-tolualdehyde, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 3-(Trifluoromethyl)benzaldehyde, Cinacalcet Impurity 23

M-Clarity™ quality level = MQ100

196878-25G 196878-5G
SIAL196878-25GEA 98.2 GBP
SIAL196878-25G SIAL196878-5G
α,α,α-Trifluoro-3-tolualdehyde, Sigma-Aldrich®
α,α,α-Trifluoro-3-tolualdehyde
Formula: C₈H₅F₃O
MW: 174.12 g/mol
Boiling Pt: 83 °C (1013 hPa)
Density: 1.3024 g/cm³ (20 °C)
MDL Number: MFCD00003373
CAS Number: 454-89-7
EINECS: 207-228-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR