Sodium trichloroacetate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: TCA-Na, Trichloroacetic acid sodium salt

M-Clarity™ quality level = MQ200

190780-100G 190780-5G
SIAL190780-100GEA 63.8 GBP
SIAL190780-100G SIAL190780-5G
Sodium trichloroacetate, Sigma-Aldrich®
Sodium trichloroacetate
Formula: C₂Cl₃NaO₂
MW: 185.37 g/mol
MDL Number: MFCD00064198
CAS Number: 650-51-1
EINECS: 211-479-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR