2-Bromothiazole, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ100
160474-25G
160474-5G
SIAL160474-25GEA
83.9
GBP
SIAL160474-25G
SIAL160474-5G
2-Bromothiazole, Sigma-Aldrich®
2-Bromothiazole
Formula:
C₃H₂BrNS MW: 164.03 g/mol Boiling Pt: 171 °C (1013 hPa) Density: 1.82 g/cm³ (20 °C) |
MDL Number:
MFCD00005316 CAS Number: 3034-53-5 EINECS: 221-229-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...