4-Formylbenzoic acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Terephthalaldehydic acid, 4-Carboxybenzaldehyde

M-Clarity™ quality level = MQ100

124915-100G 124915-10G 124915-25G
SIAL124915-100GEA 107 GBP
SIAL124915-100G SIAL124915-10G SIAL124915-25G
4-Formylbenzoic acid, Sigma-Aldrich®
4-Formylbenzoic acid
Formula: C₈H₆O₃
MW: 150.13 g/mol
Boiling Pt: 306 °C (1013 hPa)
Melting Pt: 148 °C
MDL Number: MFCD00006951
CAS Number: 619-66-9
EINECS: 210-607-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR