Safranine O

Supplier: Serva
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Safranin TSafranin Y3,7-Diamino-2,8-dimethyl-5-phenylphenazinium chlorideSafranin OGossypimine3,7-Diamino-2,8-dimethyl-5-phenylphenazin-5-ium chlorideCI 50240Basic Red 2Cotton RedSafranin ASafranin

SERA34598.01EA 0 GBP
SERA34598.01
Safranine O
Safranine O
Formula: C₂₀H₁₉ClN₄
MW: 350.85 g/mol
Density: 1.447 g/cm³ (22 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011759
CAS Number: 477-73-6
EINECS: 207-518-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR