Phthalaldehyde

Supplier: Serva
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 1,2-Phthalic dicarboxaldehyde, Phthaldialdehyde, o-Phthalaldehyde, OPA, Phthaldicarboxaldehyde, Benzene-1,2-dicarboxaldehyde

32800.01 32800.02 32800.03
SERA32800.01EA 24.47 GBP
SERA32800.01 SERA32800.02 SERA32800.03
Phthalaldehyde
Phthalaldehyde
Formula: C₈H₆O₂
MW: 134.13 g/mol
Boiling Pt: 266 °C (1013 hPa)
Melting Pt: 54…56 °C
Density: 1.13 g/cm³ (20 °C)
Flash Pt: 132 °C
MDL Number: MFCD00003335
CAS Number: 643-79-8
EINECS: 211-402-2
UN: 2923
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR