(4-Chlorophenoxy)acetic acid

Supplier: LGC Standards PROMOCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 4-CPA

PROOMM0044.02EA 829 GBP
PROOMM0044.02
(4-Chlorophenoxy)acetic acid
(4-Chlorophenoxy)acetic acid
Formula: (C₆H₄Cl)OCH₂COOH
MW: 186.59 g/mol
Boiling Pt: 308 °C (1013 hPa)
Melting Pt: 160 °C
Density: 1.53 g/cm³ (20 °C)
MDL Number: MFCD00004305
CAS Number: 122-88-3
EINECS: 204-581-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR