L(+)-Ribose
Supplier: Molekula
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
(2S,3S,4S)-2,3,4,5-Tetrahydroxypentanal
MOLE23543542-1GEA
0
GBP
MOLE23543542-1G
L(+)-Ribose
L(+)-Ribose
Formula:
C₅H₁₀O₅ MW: 150.13 g/mol Boiling Pt: 331 °C (1013 hPa) Melting Pt: 86…87 °C Storage Temperature: Ambient |
MDL Number:
MFCD00167010 CAS Number: 24259-59-4 EINECS: 246-110-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...