5-Nitrouracil
Supplier: Molekula
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
5-Nitropyrimidine-2,4(1H,3H)-dione
, Nitrouracil
MOLE11358007-100GEA
0
GBP
MOLE11358007-100G
5-Nitrouracil
5-Nitrouracil
Formula:
C₄H₃N₃O₄ MW: 157.09 g/mol Melting Pt: 300 °C Density: 1.807 g/cm³ (20 °C) |
MDL Number:
MFCD00006021 CAS Number: 611-08-5 EINECS: 210-250-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...