(Methyl)triphenylphosphonium iodide

Supplier: Molekula
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyltriphenylphosphonium iodide

MOLE19063878-100GEA 0 GBP
MOLE19063878-100G
(Methyl)triphenylphosphonium iodide
(Methyl)triphenylphosphonium iodide
Formula: C₁₉H₁₈IP
MW: 404.23 g/mol
Melting Pt: 193…194 °C
MDL Number: MFCD00066175
CAS Number: 2065-66-9
EINECS: 218-178-5
UN: 3464
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR