6-Propyl-2-thiouracil Ph. Eur.

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 4-Propyl-2-thiouracil6-Propyl-2-thioxo-2,3-dihydropyrimidin-4(1H)-one4-Hydroxy-2-mercapto-6-propylpyrimidinePropylthiouracil

8.17036.1000EA 0 GBP
8.17036.1000
6-Propyl-2-thiouracil Ph. Eur.
6-Propyl-2-thiouracil
Formula: C₇H₁₀N₂OS
MW: 170.24 g/mol
Boiling Pt: 448 °C (1013 hPa)
Melting Pt: 215…218 °C
MDL Number: MFCD00006041
CAS Number: 51-52-5
EINECS: 200-103-2
Merck Index: 12,08054

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR