D-(-)-Ribose for biochemistry, Millipore®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
(2R,3R,4R)-2,3,4,5-Tetrahydroxypentanal
M-Clarity™ quality level = MQ100
1.07605.0050EA
120
GBP
1.07605.0050
D-(-)-Ribose for biochemistry, Millipore®
D(-)-Ribose
Formula:
C₅H₁₀O₅ MW: 150.13 g/mol Boiling Pt: 331 °C (1013 hPa) Melting Pt: 90…95 °C Density: 0.8 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00135453 CAS Number: 50-69-1 EINECS: 200-059-4 Merck Index: 12,08371 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...