5-Fluorouracil
Supplier: Invivogen
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
5-Fluoropyrimidine-2,4(1H,3H)-dione
, Fluorouracil
, 5-FU
INVVSUD-5FUEA
0
GBP
INVVSUD-5FU
5-Fluorouracil
5-Fluorouracil
Formula:
C₄H₃FN₂O₂ MW: 130.08 g/mol Boiling Pt: 347 °C (1013 hPa) Melting Pt: 282…283 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006018 CAS Number: 51-21-8 EINECS: 200-085-6 UN: 2811 ADR: 6.1,III Merck Index: 12,04219 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...