1,3-Dibromo-2,4-difluorobenzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

024446-25G 024446-5G
FLUO024446-25GEA 0 GBP
FLUO024446-25G FLUO024446-5G
1,3-Dibromo-2,4-difluorobenzene
1,3-Dibromo-2,4-difluorobenzene
Formula: C₆H₂Br₂F₂
MW: 271.89 g/mol
MDL Number: MFCD07368980
CAS Number: 219926-41-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR