2-Iodo-4-(trifluoromethyl)aniline

Supplier: FLUOROCHEM LTD (CHEMPUR)
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Iodo-α,α,α-trifluoro-p-toluidine4-Amino-3-iodobenzotrifluoride4-Amino-3-iodo-α,α,α-trifluorotoluene

FLUO023846-5GEA 0 GBP
FLUO023846-25G FLUO023846-5G
2-Iodo-4-(trifluoromethyl)aniline
2-Iodo-4-(trifluoromethyl)aniline
Formula: C₇H₅F₃IN
MW: 287.02 g/mol
MDL Number: MFCD02683097
CAS Number: 163444-17-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR