1-Fluoro-3-iodo-5-nitrobenzene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
020051-1G
020051-5G
FLUO020051-1GEA
0
GBP
FLUO020051-1G
FLUO020051-5G
1-Fluoro-3-iodo-5-nitrobenzene
1-Fluoro-3-iodo-5-nitrobenzene
Formula:
C₆H₃FINO₂ MW: 267 g/mol |
MDL Number:
MFCD00007219 CAS Number: 3819-88-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...