3-(4-Methoxyphenyl)propionic acid

Supplier: FLUOROCHEM LTD (CHEMPUR)
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-(4-Methoxyphenyl)propanoic acid

FLUO018803-5GEA 0 GBP
FLUO018803-5G
3-(4-Methoxyphenyl)propionic acid
3-(4-Methoxyphenyl)propionic acid
Formula: C₁₀H₁₂O₃
MW: 180.2 g/mol
Boiling Pt: 319 °C (1013 hPa)
Melting Pt: 95…102 °C
MDL Number: MFCD00002777
CAS Number: 1929-29-9
EINECS: 217-678-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR