N-(6-Bromohexyl)phthalimide

Supplier: FLUOROCHEM LTD (CHEMPUR)
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

FLUO018278-5GEA 0 GBP
FLUO018278-5G
N-(6-Bromohexyl)phthalimide
N-(6-Bromohexyl)phthalimide
Formula: C₁₄H₁₆BrNO₂
MW: 310.19 g/mol
MDL Number: MFCD00023098
CAS Number: 24566-79-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR