2,6-Dimethylbenzoyl chloride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CFF521.2.5 017526-25G 017526-5G
CHMPCFF521.2.5EA 0 GBP
CHMPCFF521.2.5 FLUO017526-25G FLUO017526-5G
2,6-Dimethylbenzoyl chloride
2,6-Dimethylbenzoyl chloride
Formula: C₉H₉ClO
MW: 168.62 g/mol
MDL Number: MFCD00052854
CAS Number: 21900-37-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR