1-(4-Fluorobenzyl)homopiperazin

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CFF366.2.5 CFF366.5 CFF366.25 017395-2.5G 017395-5G 017395-25G
CHMPCFF366.2.5EA 0 GBP
CHMPCFF366.2.5 CHMPCFF366.5 CHMPCFF366.25 FLUO017395-2.5G FLUO017395-5G FLUO017395-25G
1-(4-Fluorobenzyl)homopiperazin
1-(4-Fluorobenzyl)homopiperazin
Formula: C₁₂H₁₇FN₂
MW: 208.28 g/mol
MDL Number: MFCD00816588
CAS Number: 76141-89-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR