2-Bromo-5-(trifluoromethyl)benzyl bromide

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Bromo-3-(bromomethyl)benzotrifluoride, 1-Bromo-2-(bromomethyl)-4-(trifluoromethyl)benzene

013792-5G 013792-1G
FLUO013792-5GEA 0 GBP
FLUO013792-5G FLUO013792-1G
2-Bromo-5-(trifluoromethyl)benzyl bromide
2-Bromo-5-(trifluoromethyl)benzyl bromide
Formula: C₈H₅Br₂F₃
MW: 317.93 g/mol
MDL Number: MFCD04973762
CAS Number: 886496-63-5
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR