3-Methylbenzofuran

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

013629-1G 013629-5G
FLUO013629-1GEA 0 GBP
FLUO013629-1G FLUO013629-5G
3-Methylbenzofuran
3-Methylbenzofuran
Formula: C₉H₈O
MW: 132.16 g/mol
Boiling Pt: 197 °C (1013 hPa)
Melting Pt: 19 °C
Density: 1.0568 g/cm³ (25 °C)
MDL Number: MFCD00205641
CAS Number: 21535-97-7
EINECS: 244-427-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR