2-Bromo-5-chlorobenzotrifluoride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Bromo-5-chlorobenzotrifluoride, 1-Bromo-4-chloro-2-(trifluoromethyl)benzene

013188-100G 013188-25G
FLUO013188-100GEA 0 GBP
FLUO013188-100G FLUO013188-25G
2-Bromo-5-chlorobenzotrifluoride
2-Bromo-5-chlorobenzotrifluoride
Formula: C₇H₃BrClF₃
MW: 259.45 g/mol
MDL Number: MFCD00010308
CAS Number: 344-65-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR