1-(2,4-Dichlorophenyl)-2-thiourea
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2,4-Dichlorophenylthiourea, 1-(2,4-Dichlorophenyl)thiourea, N-(2,4-Dichlorophenyl)thiourea
010664-5G
010664-25G
FLUO010664-5GEA
0
GBP
FLUO010664-5G
FLUO010664-25G
1-(2,4-Dichlorophenyl)-2-thiourea
1-(2,4-Dichlorophenyl)-2-thiourea
Formula:
(C₆H₃Cl₂)NC(S)NH₂ MW: 221.11 g/mol Boiling Pt: 326 °C (1013 hPa) Melting Pt: 160 °C |
MDL Number:
MFCD00022161 CAS Number: 6326-14-3 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...