3-(Diethylamino)propyl isothiocyanate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Diethylamino-1-propyl isothiocyanate, 3-(Diethylamino)propylisothiocyanate

008864-1G 008864-25G 008864-5G
FLUO008864-1GEA 0 GBP
FLUO008864-1G FLUO008864-25G FLUO008864-5G
3-(Diethylamino)propyl isothiocyanate
3-(Diethylamino)propyl isothiocyanate
Formula: C₈H₁₆N₂S
MW: 172.29 g/mol
Boiling Pt: 88…92 °C (0.1 mmHg)
Density: 0,9535 g/cm³ (20 °C)
MDL Number: MFCD00041133
CAS Number: 2626-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR