4-Bromo-2-methylphenyl isothiocyanate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

008749-5G 008749-1G
FLUO008749-5GEA 0 GBP
FLUO008749-5G FLUO008749-1G
4-Bromo-2-methylphenyl isothiocyanate
4-Bromo-2-methylphenyl isothiocyanate
Formula: C₈H₆BrNS
MW: 228.11 g/mol
Boiling Pt: 129…130 °C (3 mmHg)
Melting Pt: 50…51 °C
MDL Number: MFCD00041087
CAS Number: 19241-38-4
EINECS: 242-907-6
UN: 2923
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR