2-Chloro-5-(trifluoromethyl)benzaldehyde

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

007805-5G 007805-1G
FLUO007805-5GEA 0 GBP
FLUO007805-5G FLUO007805-1G
2-Chloro-5-(trifluoromethyl)benzaldehyde
2-Chloro-5-(trifluoromethyl)benzaldehyde
Formula: C₈H₄ClF₃O
MW: 208.57 g/mol
MDL Number: MFCD00134430
CAS Number: 82386-89-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR