2-Nitro-4-(trifluoromethoxy)aniline

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

006900-1G 006900-5G 006900-25G
FLUO006900-1GEA 0 GBP
FLUO006900-1G FLUO006900-5G FLUO006900-25G
2-Nitro-4-(trifluoromethoxy)aniline
2-Nitro-4-(trifluoromethoxy)aniline
Formula: C₇H₅F₃N₂O₃
MW: 222.12 g/mol
MDL Number: MFCD00042326
CAS Number: 2267-23-4
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR