6-Fluoro-m-anisaldehyde

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

006264-5G 006264-25G
FLUO006264-5GEA 0 GBP
FLUO006264-5G FLUO006264-25G
6-Fluoro-m-anisaldehyde
6-Fluoro-m-anisaldehyde
Formula: C₈H₇FO₂
MW: 154.14 g/mol
MDL Number: MFCD00070795
CAS Number: 105728-90-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR