1-Fluoro-4-iodo-3-nitrobenzene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
4-Fluoro-1-iodo-2-nitrobenzene, 5-Fluoro-2-iodonitrobenzene, 2-Iodo-5-fluoronitrobenzene
006246-5G
006246-1G
006246-25G
FLUO006246-5GEA
0
GBP
FLUO006246-5G
FLUO006246-1G
FLUO006246-25G
1-Fluoro-4-iodo-3-nitrobenzene
1-Fluoro-4-iodo-3-nitrobenzene
Formula:
C₆H₃FINO₂ MW: 267 g/mol Boiling Pt: 162…163 °C (35 mmHg) Melting Pt: 22.5 °C |
MDL Number:
MFCD00153169 CAS Number: 364-77-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...