3-Chloro-2,6-difluoroanisole

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Chloro-2,4-difluoro-3-methoxybenzene

005585-1G 005585-5G
FLUO005585-1GEA 0 GBP
FLUO005585-1G FLUO005585-5G
3-Chloro-2,6-difluoroanisole
3-Chloro-2,6-difluoroanisole
Formula: C₇H₅ClF₂O
MW: 178.57 g/mol
MDL Number: MFCD01631331
CAS Number: 261762-38-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR