Tetrachloro-p-benzoquinone
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
Tetrachloroquinone, 2,3,5,6-Tetrachloro-1,4-benzoquinone, 4-Chloranil, p-Chloranil, 2,3,5,6-Tetrachlorobenzo-1,4-quinone, Tetrachloro-1,4-benzoquinone
005005-25G
005005-100G
FLUO005005-25GEA
0
GBP
FLUO005005-25G
FLUO005005-100G
Tetrachloro-p-benzoquinone
Tetrachloro-p-benzoquinone
Formula:
C₆Cl₄O₂ MW: 245.88 g/mol Boiling Pt: 270 °C (1013 hPa) Melting Pt: 290…293 °C Density: 1.97 g/cm³ (20 °C) Flash Pt: >100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001594 CAS Number: 118-75-2 EINECS: 204-274-4 Merck Index: 13,02088 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...