3,5-Dichloro-4-fluorobenzotrifluoride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1,3-Dichloro-2-fluoro-5-(trifluoromethyl)benzene, 3,5-Dichloro-4-fluorobenzotrifluoride

004711-1G 004711-5G
FLUO004711-1GEA 0 GBP
FLUO004711-1G FLUO004711-5G
3,5-Dichloro-4-fluorobenzotrifluoride
3,5-Dichloro-4-fluorobenzotrifluoride
Formula: C₇H₂Cl₂F₄
MW: 232.99 g/mol
Boiling Pt: 166…168 °C (1013 hPa)
Density: 1,55 g/cm³ (20 °C)
MDL Number: MFCD00068183
CAS Number: 77227-81-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR