Bis(2-chloroethyl)ether
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-Chloroethyl ether
002891-100G
002891-500G
FLUO002891-100GEA
0
GBP
FLUO002891-100G
FLUO002891-500G
Bis(2-chloroethyl)ether
Bis(2-chloroethyl) ether
Formula:
C₄H₈Cl₂O MW: 143.01 g/mol Boiling Pt: 178 °C (1013 hPa) Melting Pt: –44 °C Density: 1.22 g/cm³ (20 °C) Flash Pt: 55 °C |
MDL Number:
MFCD00000975 CAS Number: 111-44-4 EINECS: 203-870-1 UN: 1916 ADR: 6.1,II Merck Index: 13,03091 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...