3,5-Bis(trifluoromethyl)benzonitrile
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
002099-5G
002099-25G
002099-100G
FLUO002099-5GEA
0
GBP
FLUO002099-5G
FLUO002099-25G
FLUO002099-100G
3,5-Bis(trifluoromethyl)benzonitrile
3,5-Bis(trifluoromethyl)benzonitrile
Formula:
C₉H₃F₆N MW: 239.12 g/mol Boiling Pt: 155 °C (1013 hPa) Melting Pt: 16 °C Density: 1.42 g/cm³ (20 °C) |
MDL Number:
MFCD00000379 CAS Number: 27126-93-8 EINECS: 248-240-7 UN: 3276 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...