6-Propyl-2-thiouracil
Supplier: EHRENSTORFER GMBH
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
4-Propyl-2-thiouracil
, 6-Propyl-2-thioxo-2,3-dihydropyrimidin-4(1H)-one
, 4-Hydroxy-2-mercapto-6-propylpyrimidine
, Propylthiouracil
EHERC16530500EA
99.1
GBP
EHERC16530500
6-Propyl-2-thiouracil
6-Propyl-2-thiouracil
Formula:
C₇H₁₀N₂OS MW: 170.24 g/mol Boiling Pt: 448 °C (1013 hPa) Melting Pt: 215…218 °C |
MDL Number:
MFCD00006041 CAS Number: 51-52-5 EINECS: 200-103-2 Merck Index: 12,08054 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...