(4-Chlorophenoxy)acetic acid
Supplier: EHRENSTORFER GMBH
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
4-CPA
EHERC11480000EA
135
GBP
EHERC11480000
(4-Chlorophenoxy)acetic acid
(4-Chlorophenoxy)acetic acid
Formula:
(C₆H₄Cl)OCH₂COOH MW: 186.59 g/mol Boiling Pt: 308 °C (1013 hPa) Melting Pt: 160 °C Density: 1.53 g/cm³ (20 °C) |
MDL Number:
MFCD00004305 CAS Number: 122-88-3 EINECS: 204-581-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...