2,6-Dimethoxyaniline

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CFF294.1 017336-1G
CHMPCFF294.1EA 0 GBP
CHMPCFF294.1 FLUO017336-1G
2,6-Dimethoxyaniline
2,6-Dimethoxyaniline
Formula: C₈H₁₁NO₂
MW: 153.18 g/mol
MDL Number: MFCD00053934
CAS Number: 2734-70-5
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR