4-Chloropyrocatechol

Supplier: LGC Standards PROMOCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Chloro-1,2-dihydroxybenzene4-Chlorocatechol4-Chlorobenzene-1,2-diol

PROOCILULM1701-0.1EA 0 GBP
PROOCILULM1701-0.1
4-Chloropyrocatechol
4-Chloropyrocatechol
Formula: C₆H₅ClO₂
MW: 144.56 g/mol
Boiling Pt: 267 °C (1013 hPa)
Melting Pt: 90…91 °C
MDL Number: MFCD00059614
CAS Number: 2138-22-9
EINECS: 218-381-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR