2-Acetyl-3-bromothiophene

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Acetyl-3-boromothiophene, 1-(3-Bromo-2-thienyl)-1-ethanone

9J-900.500 9J-900.1 9J-900.5 9J-900.10
BIOT9J-900.500EA 0 GBP
BIOT9J-900.500 BIOT9J-900.1 BIOT9J-900.5 BIOT9J-900.10
2-Acetyl-3-bromothiophene
2-Acetyl-3-bromothiophene
Formula: C₆H₅BrOS
MW: 205.08 g/mol
MDL Number: MFCD00794287
CAS Number: 42877-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR